Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terbium(III) fluoride, anhydrous, REacton™, 99.99% (REO)

Catalog No. p-7048676
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1121306 5 g
AA1121314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1121306 Supplier Thermo Scientific Chemicals Supplier No. 01121306
Only null left
Add to Cart

CAS: 13708-63-9 | F3Tb | 215.92 g/mol

Terbium(III) fluoride is used as a source of terbium. It is also useful in the preparation of fluoride glasses. Further, it is used in the preparation of electroluminescent thin films and luminescent zinc sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13708-63-9
Molecular Formula F3Tb
Molecular Weight (g/mol) 215.92
MDL Number MFCD00049572
InChI Key LKNRQYTYDPPUOX-UHFFFAOYSA-K
IUPAC Name terbium(3+) trifluoride
SMILES [F-].[F-].[F-].[Tb+3]

Specifications

Melting Point 1172°C
Physical Form Crystalline
Quantity 5 g
UN Number UN3288
Solubility Information Slightly soluble in mineral acids. Insoluble in water.
Formula Weight 215.92
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Terbium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 237-247-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.