Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Terbufoxon sulfoxide | 56165-57-2 | 1 MG
SDP

Catalog No. 5000417314
Encompass_Preferred

Terbufoxon sulfoxide (Terbufos oxon sulfoxide) is a metabolite of the insecticide Terbufos found in the pepper leaf matrix. This product is intended for research use only.

  • Metabolite of the insecticide Terbufos
  • For research use only
  • Molecular weight: 288.36
  • Formula: C9H21O4PS2
  • Shipping: Room temperature (continental US)
  • Storage: As per Certificate of Analysis

Catalog No. 50-004-17314 Supplier Medchemexpress LLC Supplier No. HYN80671MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Terbufoxon sulfoxide (Terbufos oxon sulfoxide) is a metabolite of the insecticide Terbufos found in the pepper leaf matrix. This product is intended for research use only.

  • Metabolite of the insecticide Terbufos
  • For research use only
  • Molecular weight: 288.36
  • Formula: C9H21O4PS2
  • Shipping: Room temperature (continental US)
  • Storage: As per Certificate of Analysis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.