Learn More
Selleck Chemical LLC Terbutaline Sulfate S4127-50mg
SDP
Catalog No. 503109947
Encompass_Preferred
Terbutaline Sulfate(Terbutaline hemisulfate) is a selective 2-adrenergic receptor agonist with IC50 of 53 nM
Catalog No. 50-310-9947 Supplier Selleck Chemical LLC Supplier No. S412750MG
Terbutaline Sulfate(Terbutaline hemisulfate) is a selective 2-adrenergic receptor agonist with IC50 of 53 nM