Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Terbutaline (sulfate) | 23031-32-5 | 100.0% | 50 MG
SDP

Catalog No. 5000409437
Encompass_Preferred

Terbutaline sulfate (Standard) is an analytical standard and orally active β2-adrenergic receptor agonist. It is intended for research and analytical applications, such as qualitative, quantitative, and methodological experiments in HPLC, GC, and MS, and can be used in asthma symptom research.

  • Analytical standard for research and analytical applications.
  • Orally active β2-adrenergic receptor agonist.
  • Active metabolite of bambuterol.
  • Can be used in asthma symptom research.
  • Suitable for HPLC, GC, and MS experiments.
  • Appears as a solid.
  • Melting/freezing point 246–248 °C.
  • Relative density 1.184 g/cm³.
  • Stable under recommended storage conditions.
  • For laboratory and manufacturing use.

Catalog No. 50-004-09437 Supplier Medchemexpress LLC Supplier No. HYB0802R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Terbutaline sulfate (Standard) is an analytical standard and orally active β2-adrenergic receptor agonist. It is intended for research and analytical applications, such as qualitative, quantitative, and methodological experiments in HPLC, GC, and MS, and can be used in asthma symptom research.

  • Analytical standard for research and analytical applications.
  • Orally active β2-adrenergic receptor agonist.
  • Active metabolite of bambuterol.
  • Can be used in asthma symptom research.
  • Suitable for HPLC, GC, and MS experiments.
  • Appears as a solid.
  • Melting/freezing point 246–248 °C.
  • Relative density 1.184 g/cm³.
  • Stable under recommended storage conditions.
  • For laboratory and manufacturing use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.