Learn More
Medchemexpress LLC Terbutaline (sulfate) | 23031-32-5 | 99.9% | 1 G
SDP
Catalog No. 5000408790
Encompass_Preferred
Terbutaline sulfate is an orally active β2-adrenergic receptor agonist and an active metabolite of bambuterol. It is used in asthma symptom research, laboratory chemicals, and for the manufacture of substances.
  • Enhances MKP-1 expression in activated mouse macrophages
  • Reduces the phosphorylation of p38 MAPK in J774 macrophages
  • Improves allodynia in ob/ob mice
  • Acts as a β adrenergic receptor agonist
  • Orally active
  • Active metabolite of bambuterol
Catalog No. 50-004-08790 Supplier Medchemexpress LLC Supplier No. HYB08021G
Terbutaline sulfate is an orally active β2-adrenergic receptor agonist and an active metabolite of bambuterol. It is used in asthma symptom research, laboratory chemicals, and for the manufacture of substances.
  • Enhances MKP-1 expression in activated mouse macrophages
  • Reduces the phosphorylation of p38 MAPK in J774 macrophages
  • Improves allodynia in ob/ob mice
  • Acts as a β adrenergic receptor agonist
  • Orally active
  • Active metabolite of bambuterol