Learn More
Medchemexpress LLC Terbutaline (sulfate) | 23031-32-5 | 99.95% | 25 MG
SDP
Catalog No. 5000450809
Encompass_Preferred
Terbutaline sulfate (Standard) is an orally active β2-adrenergic receptor agonist and an active metabolite of bambuterol. As an analytical standard, it is intended for research and analytical applications, particularly in asthma symptom research.
  • Orally active β2-adrenergic receptor agonist
  • Active metabolite of bambuterol
  • Used in asthma symptom research
  • Enhances MKP-1 expression in activated mouse macrophages
  • Reduces phosphorylation of p38 MAPK in J774 macrophages
  • Improves allodynia in ob/ob mice
  • Investigated in clinical trials for various conditions
  • Analytical standard for research and analytical applications
  • Targets the β adrenergic receptor
Catalog No. 50-004-50809 Supplier Medchemexpress LLC Supplier No. 25MGHYB0802R
Terbutaline sulfate (Standard) is an orally active β2-adrenergic receptor agonist and an active metabolite of bambuterol. As an analytical standard, it is intended for research and analytical applications, particularly in asthma symptom research.
  • Orally active β2-adrenergic receptor agonist
  • Active metabolite of bambuterol
  • Used in asthma symptom research
  • Enhances MKP-1 expression in activated mouse macrophages
  • Reduces phosphorylation of p38 MAPK in J774 macrophages
  • Improves allodynia in ob/ob mice
  • Investigated in clinical trials for various conditions
  • Analytical standard for research and analytical applications
  • Targets the β adrenergic receptor