Learn More
Cayman Chemical TerbuthylazIn 10mg
SDP
Catalog No. 502950736
Encompass_Preferred
A triazine herbicide; active against a variety of broadleaf and grass weeds in combination with S-metolachlor; has been found in surface groundwater and soil samples
Catalog No. 50-295-0736 Supplier Cayman Chemical Supplier No. 3419210 MG
A triazine herbicide; active against a variety of broadleaf and grass weeds in combination with S-metolachlor; has been found in surface groundwater and soil samples