Learn More
Cayman Chemical TerbuthylazIn 50mg
SDP
Catalog No. 502946738
Encompass_Preferred
A triazine herbicide; active against a variety of broadleaf and grass weeds in combination with S-metolachlor; has been found in surface groundwater and soil samples
Catalog No. 50-294-6738 Supplier Cayman Chemical Supplier No. 3419250 MG
A triazine herbicide; active against a variety of broadleaf and grass weeds in combination with S-metolachlor; has been found in surface groundwater and soil samples