Learn More
Medchemexpress LLC HY-B1790 100mg Medchemexpress, Terconazole CAS:67915-31-5 Purity:>98%
SDP
Catalog No. 501933010
Encompass_Preferred
Medchemexpress, HY-B1790 100mg Terconazole CAS:67915-31-5 Terconazole is a broad-spectrum antifungal medication for the treatment of vaginal yeast infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3010 Supplier Medchemexpress LLC Supplier No. HYB1790100MG
Medchemexpress, HY-B1790 100mg Terconazole CAS:67915-31-5 Terconazole is a broad-spectrum antifungal medication for the treatment of vaginal yeast infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.