Learn More
Medchemexpress LLC Terephthalaldehyde | 623-27-8 | MFCD00006949 | 99.9% | 134.13 | C8H6O2 | 250g
SDP
Catalog No. 5000268278
Encompass_Preferred
Terephthalaldehyde is a cross-linking agent Terephthalaldehyde can copolymerize with chitosan and acrylic acid to form hydrogels that selectively adsorb anionic dyes Terephthalaldehyde can be used as a drug intermediate to synthesize a series of novel benzothiazole and benzotriazolopyrimidinone heterocyclic compounds[1][2]
Catalog No. 50-002-68278 Supplier Medchemexpress LLC Supplier No. HYY0425250G
Terephthalaldehyde is a cross-linking agent Terephthalaldehyde can copolymerize with chitosan and acrylic acid to form hydrogels that selectively adsorb anionic dyes Terephthalaldehyde can be used as a drug intermediate to synthesize a series of novel benzothiazole and benzotriazolopyrimidinone heterocyclic compounds[1][2]