Learn More
Medchemexpress LLC Terephthalaldehyde | 623-27-8 | MFCD00006949 | 99.9% | 134.13 g/mol | C8H6O2 | 500g
SDP
Catalog No. 5000268227
Encompass_Preferred
Terephthalaldehyde is a cross-linking agent Terephthalaldehyde can copolymerize with chitosan and acrylic acid to form hydrogels that selectively adsorb anionic dyes Terephthalaldehyde can be used as a drug intermediate to synthesize a series of novel benzothiazole and benzotriazolopyrimidinone heterocyclic compounds[1][2]
Catalog No. 50-002-68227 Supplier Medchemexpress LLC Supplier No. HYY0425500G
Terephthalaldehyde is a cross-linking agent Terephthalaldehyde can copolymerize with chitosan and acrylic acid to form hydrogels that selectively adsorb anionic dyes Terephthalaldehyde can be used as a drug intermediate to synthesize a series of novel benzothiazole and benzotriazolopyrimidinone heterocyclic compounds[1][2]