Learn More
Medchemexpress LLC Terephthalic acid | 100-21-0 | 99.99% | 166.13 | 100 MG
SDP
Catalog No. 5000452545
Encompass_Preferred
Terephthalic acid is an isomer and precursor of polyester PET, widely used in textiles and plastics. It is also utilized in biological research for developing skin tumor models. This compound promotes cell proliferation and can induce skin tumor formation by interfering with the p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway.
  • Isomer and precursor of polyester PET
  • Used in clothing and plastics
  • Can be used to build skin tumor models
  • Promotes excessive cell proliferation
  • Induces skin tumor formation
  • May lead to bladder stone formation at higher doses
Catalog No. 50-004-52545 Supplier Medchemexpress LLC Supplier No. HYW010098R100MG
Add to Cart
Add to Cart
Terephthalic acid is an isomer and precursor of polyester PET, widely used in textiles and plastics. It is also utilized in biological research for developing skin tumor models. This compound promotes cell proliferation and can induce skin tumor formation by interfering with the p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway.
  • Isomer and precursor of polyester PET
  • Used in clothing and plastics
  • Can be used to build skin tumor models
  • Promotes excessive cell proliferation
  • Induces skin tumor formation
  • May lead to bladder stone formation at higher doses