Learn More
Medchemexpress LLC Terephthalic acid | 100-21-0 | 99.99% | 166.13 | 25 MG
SDP
Catalog No. 5000366726
Encompass_Preferred
Terephthalic acid is an isomer and a precursor of polyester PET, which can be used to make clothing and plastics. It can also be used to build skin tumor models.
  • Isomer and precursor of polyester PET.
  • Can be used to build skin tumor models.
  • Can promote excessive cell proliferation and induce skin tumor formation by disrupting the p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway.
  • In animal modeling, it can create skin tumor models.
  • Metabolized in the body to form calcium terephthalate or calcium hydrogen terephthalate, leading to oversaturation in the urine and promoting the formation of bladder stones.
Catalog No. 50-003-66726 Supplier Medchemexpress LLC Supplier No. 25MGHYW010098R
Terephthalic acid is an isomer and a precursor of polyester PET, which can be used to make clothing and plastics. It can also be used to build skin tumor models.
  • Isomer and precursor of polyester PET.
  • Can be used to build skin tumor models.
  • Can promote excessive cell proliferation and induce skin tumor formation by disrupting the p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway.
  • In animal modeling, it can create skin tumor models.
  • Metabolized in the body to form calcium terephthalate or calcium hydrogen terephthalate, leading to oversaturation in the urine and promoting the formation of bladder stones.