Learn More
Medchemexpress LLC Terephthalic acid | 100-21-0 | 99.99% | 166.13 | 50 G
SDP
Catalog No. 5000453538
Encompass_Preferred
Terephthalic acid is an isomer and a precursor of polyester PET, used in clothing and plastics. It can also be utilized to build skin tumor models and promote excessive cell proliferation by disrupting the p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway. In vivo, it can lead to bladder stone formation in animal models, particularly in weanling mice at higher doses.
  • Isomer and precursor of polyester PET
  • Used to build skin tumor models
  • Promotes excessive cell proliferation
  • Induces skin tumor formation
  • Metabolized to form calcium terephthalate, potentially leading to bladder stone formation
Catalog No. 50-004-53538 Supplier Medchemexpress LLC Supplier No. HYW01009850G
Terephthalic acid is an isomer and a precursor of polyester PET, used in clothing and plastics. It can also be utilized to build skin tumor models and promote excessive cell proliferation by disrupting the p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway. In vivo, it can lead to bladder stone formation in animal models, particularly in weanling mice at higher doses.
  • Isomer and precursor of polyester PET
  • Used to build skin tumor models
  • Promotes excessive cell proliferation
  • Induces skin tumor formation
  • Metabolized to form calcium terephthalate, potentially leading to bladder stone formation