Learn More
Medchemexpress LLC Terephthalic acid-13C2 | 121191-53-5 | 99.6% | 168.12 | 1 MG
SDP
Catalog No. 5000453981
Encompass_Preferred
Terephthalic acid-13C2 is the 13C-labeled version of Terephthalic acid, which is an isomer and a precursor to polyester PET used in clothing and plastics. It can also be utilized to create skin tumor models. This compound is intended for research use only and is not sold to patients.
  • Can be used as a tracer and as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes of carbon (13C), which are incorporated into drug molecules for quantitation during drug development.
Catalog No. 50-004-53981 Supplier Medchemexpress LLC Supplier No. HYW010098S11MG
Terephthalic acid-13C2 is the 13C-labeled version of Terephthalic acid, which is an isomer and a precursor to polyester PET used in clothing and plastics. It can also be utilized to create skin tumor models. This compound is intended for research use only and is not sold to patients.
  • Can be used as a tracer and as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes of carbon (13C), which are incorporated into drug molecules for quantitation during drug development.