Learn More
Medchemexpress LLC Terephthalic acid-13C2 | 121191-53-5 | 99.6% | 168.12 | 10 MG
SDP
Catalog No. 5000453980
Encompass_Preferred
Terephthalic acid-13C2 is a 13C-labeled isomer of Terephthalic acid, a precursor of polyester PET. It is utilized in the production of clothing and plastics and can also be employed in building skin tumor models.
  • Used as a tracer.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-53980 Supplier Medchemexpress LLC Supplier No. HYW010098S110MG
Add to Cart
Add to Cart
Terephthalic acid-13C2 is a 13C-labeled isomer of Terephthalic acid, a precursor of polyester PET. It is utilized in the production of clothing and plastics and can also be employed in building skin tumor models.
  • Used as a tracer.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.