Learn More
Medchemexpress LLC Terephthalic acid-13C2 | 121191-53-5 | 99.6% | 168.12 | 100 MG
SDP
Catalog No. 5000453540
Encompass_Preferred
Terephthalic acid-13C2 is the 13C-labeled version of Terephthalic acid, an isomer and precursor of polyester PET. It can also be used to create skin tumor models.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during drug development
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-53540 Supplier Medchemexpress LLC Supplier No. HYW010098S1100MG
Terephthalic acid-13C2 is the 13C-labeled version of Terephthalic acid, an isomer and precursor of polyester PET. It can also be used to create skin tumor models.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during drug development
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs