Learn More
Medchemexpress LLC Terephthalic acid-13C2 | 121191-53-5 | 99.6% | 168.12 | 25 MG
SDP
Catalog No. 5000366738
Encompass_Preferred
Terephthalic acid-13C2 is the 13C-labeled version of Terephthalic acid. Terephthalic acid is an isomer and a precursor of polyester PET, which is used in clothing and plastics. It can also be utilized to create skin tumor models.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as carbon-13 (13C), are incorporated into drug molecules primarily as tracers for quantification during drug development.
Catalog No. 50-003-66738 Supplier Medchemexpress LLC Supplier No. 25MGHYW010098S1
Add to Cart
Add to Cart
Terephthalic acid-13C2 is the 13C-labeled version of Terephthalic acid. Terephthalic acid is an isomer and a precursor of polyester PET, which is used in clothing and plastics. It can also be utilized to create skin tumor models.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as carbon-13 (13C), are incorporated into drug molecules primarily as tracers for quantification during drug development.