Learn More
Medchemexpress LLC Terephthalic acid-13C2 | 121191-53-5 | 99.6% | 168.12 | 50 MG
SDP
Catalog No. 5000453539
Encompass_Preferred
Terephthalic acid-13C2 is the 13C-labeled version of Terephthalic acid. Terephthalic acid is an isomer and a precursor of polyester PET, a material used for clothing and plastics. It can also be used to build skin tumor models.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS
  • Stable heavy isotopes like carbon-13 have been incorporated into drug molecules for quantitation during drug development
Catalog No. 50-004-53539 Supplier Medchemexpress LLC Supplier No. HYW010098S150MG
Terephthalic acid-13C2 is the 13C-labeled version of Terephthalic acid. Terephthalic acid is an isomer and a precursor of polyester PET, a material used for clothing and plastics. It can also be used to build skin tumor models.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS
  • Stable heavy isotopes like carbon-13 have been incorporated into drug molecules for quantitation during drug development