Learn More
Selleck Chemical LLC Terephthalic acid
SDP
Catalog No. 5000003228
Encompass_Preferred
Terephthalic acid (para-Phthalic acid 1 4-Benzenedioic acid) a raw material for polyethylene terephthalate (PET) production is one of the most important chemicals in petrochemical industry Terephthalic acid can be used to induce animal models of Bladder Urolithiasis
Catalog No. 50-000-03228 Supplier Selleck Chemical LLC Supplier No. S625110MM/1ML
Terephthalic acid (para-Phthalic acid 1 4-Benzenedioic acid) a raw material for polyethylene terephthalate (PET) production is one of the most important chemicals in petrochemical industry Terephthalic acid can be used to induce animal models of Bladder Urolithiasis