Learn More
Selleck Chemical LLC Terephthalic acid
SDP
Supplier:  Selleck Chemical LLC S62511G
Encompass_Preferred
Terephthalic acid (para-Phthalic acid 1 4-Benzenedioic acid) a raw material for polyethylene terephthalate (PET) production is one of the most important chemicals in petrochemical industry Terephthalic acid can be used to induce animal models of Bladder Urolithiasis
Catalog No. 50-000-03777