Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terephthalic acid, ≥98.0% (GC); 97.5-102.5% (T), MilliporeSigma™ Supelco™

Catalog No. 111012270 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2270 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2270 Supplier MilliporeSigma Supelco Supplier No. 40818100MG
Add to Cart
Add to Cart

Specifications

Ignition Point 925°F
CAS 100-21-0
Vapor Pressure <0.01 mmHg (20°C)
Physical Form Neat
Linear Formula C6H4-1,4-(CO2 H)2
MDL Number MFCD00002558
Melting Point >300°C
Synonym Benzene-1,4-dicarboxylic acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 166.13
Percent Purity ≥98.0% (GC); 97.5 to 102.5% (T)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.