Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terephthalic acid, 99+%, ACROS Organics™

Click to view available options
Quantity:
250g
500g
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic bottle
100-21-0
C8H6O4
166.13
MFCD00002558
KKEYFWRCBNTPAC-UHFFFAOYSA-N
p-phthalic acid,1,4-benzenedicarboxylic acid,benzene-1,4-dicarboxylic acid,p-dicarboxybenzene,p-benzenedicarboxylic acid,p-carboxybenzoic acid,acide terephtalique,para-phthalic acid,tephthol,1,4-dicarboxybenzene
7489
CHEBI:15702
terephthalic acid
C1=CC(=CC=C1C(=O)O)C(=O)O
100-21-0
C8H6O4
166.13
MFCD00002558
KKEYFWRCBNTPAC-UHFFFAOYSA-N
p-phthalic acid,1,4-benzenedicarboxylic acid,benzene-1,4-dicarboxylic acid,p-dicarboxybenzene,p-benzenedicarboxylic acid,p-carboxybenzoic acid,acide terephtalique,para-phthalic acid,tephthol,1,4-dicarboxybenzene
7489
CHEBI:15702
terephthalic acid
C1=CC(=CC=C1C(=O)O)C(=O)O
Packaging Plastic bottle
Sulfated Ash 0.001% max.
Melting Point >300.0°C
Infrared Spectrum Authentic
Water 0.2% max.
Quantity 250g
Color White
Sublimation Point 0402.0°C
Assay Percent Range 99+%
Beilstein 09,841
Merck Index 15,9305
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkalies, slightly soluble in alcohol, insoluble in chloroform, ether and acetic acid
Formula Weight 166.13
Physical Form Crystalline Powder
Percent Purity 99+%
Chemical Name or Material Terephthalic acid
Show More Show Less

EINECSNumber : 202-830-