Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Terephthalic acid disodium salt | 10028-70-3 | MFCD00013137 | 99.9% | 210.09 g/mol | C8H4Na2O4 | 5 G
SDP

Catalog No. 5000280712
Encompass_Preferred

Disodium terephthalate is the disodium salt of terephthalic acid, used as a chemical building block and biochemical reagent. It serves as a precursor in polyester (PET) production and is used in laboratory research applications, including formulation of skin tumor models. The material is provided as a solid for bench-scale experimental work.

  • High purity suitable for analytical and research applications.
  • Molecular weight 210.09 g/mol; formula C8H4Na2O4.
  • Soluble in water (~70 mg/mL); ultrasonic agitation may be required.
  • Supplied in small pack sizes convenient for laboratory use.
  • Useful as a PET precursor and reagent in biochemical assays.

Catalog No. 50-002-80712 Supplier Medchemexpress LLC Supplier No. HYW2501275G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disodium terephthalate is the disodium salt of terephthalic acid, used as a chemical building block and biochemical reagent. It serves as a precursor in polyester (PET) production and is used in laboratory research applications, including formulation of skin tumor models. The material is provided as a solid for bench-scale experimental work.

  • High purity suitable for analytical and research applications.
  • Molecular weight 210.09 g/mol; formula C8H4Na2O4.
  • Soluble in water (~70 mg/mL); ultrasonic agitation may be required.
  • Supplied in small pack sizes convenient for laboratory use.
  • Useful as a PET precursor and reagent in biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.