Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Terephthalic acid (Standard) | 100-21-0 | 99.9% | 166.13 | 10 MG
SDP

Catalog No. 5000452546
Encompass_Preferred

Terephthalic acid (Standard) is the analytical standard of Terephthalic acid. It is intended for research and analytical applications. Terephthalic acid is an isomer and a precursor of polyester PET, which can be used to make clothing and plastics. It can also be used to build skin tumor models.

  • Analytical standard, used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Please store the product under the recommended conditions in the Certificate of Analysis

Catalog No. 50-004-52546 Supplier Medchemexpress LLC Supplier No. HYW010098R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Terephthalic acid (Standard) is the analytical standard of Terephthalic acid. It is intended for research and analytical applications. Terephthalic acid is an isomer and a precursor of polyester PET, which can be used to make clothing and plastics. It can also be used to build skin tumor models.

  • Analytical standard, used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Please store the product under the recommended conditions in the Certificate of Analysis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.