Learn More
Medchemexpress LLC Terephthalic acid (standard) | 100-21-0 | >99.9% | 166.13 | 50 MG
SDP
Catalog No. 5000453979
Encompass_Preferred
Terephthalic acid (Standard) is an analytical standard intended for research and analytical applications. It is an isomer and a precursor of polyester PET, which is used to make clothing and plastics. This product can also be used to build skin tumor models.
  • Analytical standard for research and analytical applications.
  • Reference standard for assays.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Precursor of polyester PET.
  • Can be used to build skin tumor models.
Catalog No. 50-004-53979 Supplier Medchemexpress LLC Supplier No. HYW010098R50MG
Terephthalic acid (Standard) is an analytical standard intended for research and analytical applications. It is an isomer and a precursor of polyester PET, which is used to make clothing and plastics. This product can also be used to build skin tumor models.
  • Analytical standard for research and analytical applications.
  • Reference standard for assays.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Precursor of polyester PET.
  • Can be used to build skin tumor models.