Learn More
Medchemexpress LLC Terephthalic acid (Standard) | 100-21-0 | 99.99% | 166.13 | 25 G
SDP
Catalog No. 5000366771
Encompass_Preferred
Terephthalic acid is an isomer and a precursor of polyester PET, used to make clothing and plastics. It can also be utilized to build skin tumor models by promoting excessive cell proliferation and disrupting the p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway. This product is for research use only.
  • Used in animal modeling to create skin tumor models
  • Metabolized in the body to form calcium terephthalate or calcium hydrogen terephthalate
  • Promotes formation of bladder stones
  • Promotes excessive cell proliferation
  • Induces skin tumor formation by disrupting p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway
  • Available in solid form
  • Soluble in DMSO
  • Stable under recommended storage conditions
  • Room temperature shipping
Catalog No. 50-003-66771 Supplier Medchemexpress LLC Supplier No. 25GHYW010098
Terephthalic acid is an isomer and a precursor of polyester PET, used to make clothing and plastics. It can also be utilized to build skin tumor models by promoting excessive cell proliferation and disrupting the p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway. This product is for research use only.
  • Used in animal modeling to create skin tumor models
  • Metabolized in the body to form calcium terephthalate or calcium hydrogen terephthalate
  • Promotes formation of bladder stones
  • Promotes excessive cell proliferation
  • Induces skin tumor formation by disrupting p16-cyclin D1/Cdk4-Rb cell cycle regulatory pathway
  • Available in solid form
  • Soluble in DMSO
  • Stable under recommended storage conditions
  • Room temperature shipping