Learn More
Medchemexpress LLC Terevalefim (ANG-3777) | 1070881-42-3 | 99.9% | 176.24 g/mol | C9H8N2S | 5 MG
SDP
Catalog No. 5000223372
Encompass_Preferred
Terevalefim (ANG-3777) is a small-molecule hepatocyte growth factor (HGF) mimetic that selectively activates the c-Met receptor. It reduces apoptosis, promotes cell proliferation, and has shown efficacy in preclinical models supporting organ repair and function. Intended for research use only.
  • Selectively activates the c-Met receptor.
  • Mimics hepatocyte growth factor activity to reduce apoptosis and promote proliferation.
  • Demonstrated organ repair and functional improvement in animal models.
  • Available as solid and as a DMSO solution for flexible experimental use.
  • High purity suitable for research applications.
Catalog No. 50-002-23372 Supplier Medchemexpress LLC Supplier No. HY1374555MG
Terevalefim (ANG-3777) is a small-molecule hepatocyte growth factor (HGF) mimetic that selectively activates the c-Met receptor. It reduces apoptosis, promotes cell proliferation, and has shown efficacy in preclinical models supporting organ repair and function. Intended for research use only.
  • Selectively activates the c-Met receptor.
  • Mimics hepatocyte growth factor activity to reduce apoptosis and promote proliferation.
  • Demonstrated organ repair and functional improvement in animal models.
  • Available as solid and as a DMSO solution for flexible experimental use.
  • High purity suitable for research applications.