Learn More
Aviva Systems Biology Corp TERF2 Antibody - C-terminal region (ARP30228_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30228_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TERF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TERF2. Alternate names: TRBF2, TRF2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52821