Learn More
Aviva Systems Biology Corp TERF2IP Antibody - middle region (ARP57302_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57302_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TERF2IP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TERF2IP. Alternate names: DRIP5, RAP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53861