Learn More
Medchemexpress LLC Terfenadine | 50679-08-8 | MFCD00079622 | 99.9% | 471.67 g·mol-1 | C32H41NO2 | 10 MG
SDP
Catalog No. 5000298202
Encompass_Preferred
Terfenadine is an analytical reference standard used for research and analytical applications, including HPLC, GC, and MS. It is a histamine H1 receptor antagonist and a known hERG channel blocker, commonly used in method development, qualitative and quantitative assays, and stability studies.
  • Analytical standard suitable for HPLC, GC, and MS.
  • High purity (99.9%), suitable for analytical quantification.
  • Useful for method development, calibration, and stability testing.
  • Supplied as a 10 MG quantity for laboratory use.
  • Compatible with common analytical workflows and instruments.
Catalog No. 50-002-98202 Supplier Medchemexpress LLC Supplier No. HYB1193R10MG
Terfenadine is an analytical reference standard used for research and analytical applications, including HPLC, GC, and MS. It is a histamine H1 receptor antagonist and a known hERG channel blocker, commonly used in method development, qualitative and quantitative assays, and stability studies.
  • Analytical standard suitable for HPLC, GC, and MS.
  • High purity (99.9%), suitable for analytical quantification.
  • Useful for method development, calibration, and stability testing.
  • Supplied as a 10 MG quantity for laboratory use.
  • Compatible with common analytical workflows and instruments.