Learn More
Research Products International Corp Tergazyme Enzyme Powder Detergent, 4 Pounds
SDP
Catalog No. 50476437
Encompass
Scientifically formulated anionic powder detergent with protease enzyme for manual and ultrasonic cleaning. Excellent formula for removal of proteinaceaous soils, tissue, blood, and bodily fluids from glass, metal, plastic, ceramic, porcelain, rubber and hard surface materials. Cleans effectively and leaves no residues. Ideal cleaning agent in reverse osmosis and ultra-filtration systems.
Catalog No. 50-476-437 Supplier Research Products International Corp Supplier No. 114016
Scientifically formulated anionic powder detergent with protease enzyme for manual and ultrasonic cleaning. Excellent formula for removal of proteinaceaous soils, tissue, blood, and bodily fluids from glass, metal, plastic, ceramic, porcelain, rubber and hard surface materials. Cleans effectively and leaves no residues. Ideal cleaning agent in reverse osmosis and ultra-filtration systems.