Learn More
Research Products International Corp Tergajet Powder Detergent, 4 Pounds
SDP
Catalog No. 50998425
Encompass
Tergajet is a scientifically formulated nonionic, low foaming, phosphate-free powder detergent. Removes soil, grit, and blood from glass, metal plastic, rubber and hard surface materials. Ideal for use in labware washers, manual and ultrasonic cleaning.
Catalog No. 50-998-425 Supplier Research Products International Corp Supplier No. 114015
Tergajet is a scientifically formulated nonionic, low foaming, phosphate-free powder detergent. Removes soil, grit, and blood from glass, metal plastic, rubber and hard surface materials. Ideal for use in labware washers, manual and ultrasonic cleaning.