Learn More
Andwin Scientific TERGAJET DTRGENT 4LB--QTY9--
SDP
Catalog No. 50997509
Encompass_Preferred
The Tergajet low-foaming powdered detergent can be utilized to santize water testing labwares, phosphate-sensitive lab equipments, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks, and reactors. The detergent can eliminate different particles, such as soil, grit, tars, grease, blood, tissue, and several more. It is sold in a quantity of 4 lbs.
Catalog No. 50-997-509 Supplier Andwin Scientific Supplier No. 6053401
The Tergajet low-foaming powdered detergent can be utilized to santize water testing labwares, phosphate-sensitive lab equipments, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks, and reactors. The detergent can eliminate different particles, such as soil, grit, tars, grease, blood, tissue, and several more. It is sold in a quantity of 4 lbs.