Learn More
Electron Microscopy Sciences Tergajet Low Foaming Phosphate Free Powder 4 LB
SDP
Catalog No. 501898970
Encompass_Preferred
Non-ionic, low foaming phosphate free powdered enzyme cleaner for use in labware washers, manual and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porselain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues. Passes inhibitor residue test. Dilute 1:200 and it has a pH 11.5.
Catalog No. 50-189-8970 Supplier Electron Microscopy Sciences Supplier No. 6053401
Non-ionic, low foaming phosphate free powdered enzyme cleaner for use in labware washers, manual and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porselain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues. Passes inhibitor residue test. Dilute 1:200 and it has a pH 11.5.