Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tergitol(R) 15-S-5, Surfactant, Spectrum™ Chemical
SDP

Catalog No. p-7158357
Click to view available options
Volume (Metric):
500 mL
4 L

T1255, 84133-50-6, C12-14H25-29O(CH2CH2O)xH

Spectrum™ Chemical Tergitol(R) 15-S-5, Surfactant is a nonionic Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints & coatings.

Specifications

pH 5.5 to 7.5
Composition C12-14-H25-29-O(CH2-CCH2)xH
Volume (Metric) 500 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.