Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tergitol(R) 15-S-9, Surfactant, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Volume (Metric):
500 mL
4 L
Description
Spectrum™ Chemical Tergitol(R) 15-S-9, Surfactant is a nonionic Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints & coatings.
Specifications
Specifications
| pH | 5.5 to 7.5 |
| Composition | C12-14H25-29O[CH2CH2O]xH |
| Volume (Metric) | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction