Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tergitol(R) 15-S-9, Surfactant, Spectrum™ Chemical
SDP

Catalog No. 18607261 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Volume (Metric):
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
18-607-261 4 L
18-607-260 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-261 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T12614LTGL

T1261, 84133-50-6, C12-14H25-29O[CH2CH2O]xH

Spectrum™ Chemical Tergitol(R) 15-S-9, Surfactant is a nonionic Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints & coatings.

Specifications

pH 5.5 to 7.5
Composition C12-14H25-29O[CH2CH2O]xH
Volume (Metric) 4 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.