Learn More
Description
Spectrum™ Chemical Tergitol NP-4, Surfactant is a Nonylphenol Ethoxylate surfactant. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners and degreasers, prewash spotters and oil field chemicals.
Specifications
Specifications
| CAS Max % | 1 |
| pH | 5.0 to 8.0 |
| Quantity | 100 mL |
| Packaging | Amber Glass Bottle |
| Water | 0.005 |
| Grade | Reagent |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
