Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tergitol(R) NP-9, Surfactant, Spectrum™ Chemical
SDP

Catalog No. 18607280 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-280 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-280 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1285500ML

missing translation for 'validMainframeMsg'

T1285, 127087-87-0, C15H24O(C2H4O)x

Spectrum™ Chemical Tergitol NP-9, Surfactant is a Nonylphenol Ethoxylate based surfactant. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners and detergents, prewash spotters, paints and coatings, dust control and metalworking fluids.

Chemical Identifiers

CAS 127087-87-0

Specifications

CAS Max % 1
pH 5.0 to 8.0
Quantity 500 mL
Packaging Amber Glass Bottle
Specific Gravity 1.042-1.062
Water 0.003
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.