Learn More
Apexbio Technology LLC Teriflunomide, 10mg. Cas: 108605-62-5 MFCD: MFCD00910058. Inhibitor of dihydroorotate dehydrogenase
SDP
Supplier:  Apexbio Technology LLC B185010
Encompass_Preferred
Teriflunomide (also known as A77 1726), the active metabolite of an approved antirheumatic drug Leflunomide, is an emerging oral therapy for multiple sclerosis (MS). Teriflunomide has demonstrated anti-inflammatory, antiproliferative and immunosuppressive Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5287