Learn More
Apexbio Technology LLC Teriflunomide 108605-62-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063721
Encompass_Preferred
Teriflunomide (CAS 108605-62-5) the active metabolite of the antirheumatic agent Leflunomide acts primarily by selectively inhibiting dihydroorotate dehydrogenase (DHODH) a mitochondrial enzyme critical for pyrimidine nucleotide synthesis By restricting pyrimidine production teriflunomide exerts antiproliferative effects on activated lymphocytes resulting in immunosuppressive and anti-inflammatory activity Due to these properties teriflunomide is extensively studied as an oral therapeutic candidate in multiple sclerosis research addressing autoimmune-mediated disorders
Catalog No. 50-000-63721 Supplier Apexbio Technology LLC Supplier No. B18505.1
Teriflunomide (CAS 108605-62-5) the active metabolite of the antirheumatic agent Leflunomide acts primarily by selectively inhibiting dihydroorotate dehydrogenase (DHODH) a mitochondrial enzyme critical for pyrimidine nucleotide synthesis By restricting pyrimidine production teriflunomide exerts antiproliferative effects on activated lymphocytes resulting in immunosuppressive and anti-inflammatory activity Due to these properties teriflunomide is extensively studied as an oral therapeutic candidate in multiple sclerosis research addressing autoimmune-mediated disorders