Learn More
Selleck Chemical LLC Teriflunomide 50mg 108605-62-5 A77 1726, HMR-1726
SDP
Supplier:  Selleck Chemical LLC S416950MG
Encompass_Preferred
Teriflunomide (A77 1726, HMR-1726) is the active metabolite of leflunomide, inhibiting pyrimidine de novo synthesis by blocking the enzyme dihydroorotate dehydrogenase, used as an immunomodulatory agent. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0889697