Learn More
Medchemexpress LLC HY-15405 50mg , Teriflunomide CAS:163451-81-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1540550MG
Encompass_Preferred
HY-15405 50mg Teriflunomide CAS: 163451-81-8 Teriflunomide is the active metabolite of leflunomide, an approved therapy for rheumatoid arthritis. It inhibits pyrimidine synthesis and therefore potently decreases T cell and B cell proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0159