Learn More
Medchemexpress LLC Teriflunomide-d4 | 1185240-22-5 | 99.98% | 274.23 | 50 MG
SDP
Catalog No. 5000344057
Encompass_Preferred
Teriflunomide-d4 is deuterium labeled Teriflunomide. It is intended for research use only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-44057 Supplier Medchemexpress LLC Supplier No. HY110159S1MG
Teriflunomide-d4 is deuterium labeled Teriflunomide. It is intended for research use only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.