Learn More
TARGETMOL CHEMICALS INC Teriparatide acetate 5MG
SDP
Catalog No. 502879477
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Teriparatide acetate (Forteo) is a PHT agonist, with an IC50 of 2 nM in HEK293 cells.Teriparatide acetate is a recombinant form of parathyroid hormone. It is an effective anabolic (i.e., bone growing) agent used to treat some forms of osteoporosis. Intermittent use of Teriparatide acetate activates osteoblasts more than osteoclasts, which leads to an overall increase in bone. Purity 99.86%
Catalog No. 50-287-9477 Supplier TARGETMOL CHEMICALS INC Supplier No. T214605MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Teriparatide acetate (Forteo) is a PHT agonist, with an IC50 of 2 nM in HEK293 cells.Teriparatide acetate is a recombinant form of parathyroid hormone. It is an effective anabolic (i.e., bone growing) agent used to treat some forms of osteoporosis. Intermittent use of Teriparatide acetate activates osteoblasts more than osteoclasts, which leads to an overall increase in bone. Purity 99.86%