Learn More
Eurofins Discoverx cAMP Hunter Teriparatide Bioassay Kit, 10pl. (95-0118Y2-00058)
Catalog No. 502003284
Encompass
The cAMP Hunter Teriparatide Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses Teriparatide based activation of PTHR GPCR activity via detection of levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTHR, a naturally Gs coupled GPCR. Upon activation of PTHR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-3284 Supplier Eurofins Discoverx Supplier No. 950118Y200058
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter Teriparatide Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses Teriparatide based activation of PTHR GPCR activity via detection of levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTHR, a naturally Gs coupled GPCR. Upon activation of PTHR, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.