Learn More
Cayman Chemical Terlipressin (acetate), Purity: ≥95%, CAS: 914453-96-6, 50 mg
SDP
Catalog No. 502937363
Encompass_Preferred
A prodrug form of lysipressin; a partial agonist of the vasopressin V1A receptor (Ki = 0.85 µM); an agonist of vasopressin V1B and V2 receptors (Kis = 1.11 and 1.58 µM, respectively); increases MAP and decreases mortality in a rat model of uncontrolled hemorrhagic shock at 15 µg/kg; improves vascular reactivity and increases survival time in a rat model of cecal ligation and puncture-induced septic shock and a rabbit model of LPS-induced septic shock at 2.6 µg/kg per hour in combination with norepinephrine; increases MAP in a rat model of liver cirrhosis
Catalog No. 50-293-7363 Supplier Cayman Chemical Supplier No. 2702750 MG
A prodrug form of lysipressin; a partial agonist of the vasopressin V1A receptor (Ki = 0.85 µM); an agonist of vasopressin V1B and V2 receptors (Kis = 1.11 and 1.58 µM, respectively); increases MAP and decreases mortality in a rat model of uncontrolled hemorrhagic shock at 15 µg/kg; improves vascular reactivity and increases survival time in a rat model of cecal ligation and puncture-induced septic shock and a rabbit model of LPS-induced septic shock at 2.6 µg/kg per hour in combination with norepinephrine; increases MAP in a rat model of liver cirrhosis