Learn More
TARGETMOL CHEMICALS INC Terlipressin Acetate 25MG
SDP
Catalog No. 502876393
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Terlipressin Acetate, a highly selective vasopressin V1 receptor agonist, on oxygen and glucose deprivation/re-oxygenation (OGD/R)-induced damage in intestinal epithelial cells (IEC-6).Terlipressin acetate exerts anti-inflammatory and anti-oxidative effects. Terlipressin acetate has the potential for hepatorenal syndrome and norepinephrine-resistant septic shock research. Purity 99.69%
Catalog No. 50-287-6393 Supplier TARGETMOL CHEMICALS INC Supplier No. T13920L25MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Terlipressin Acetate, a highly selective vasopressin V1 receptor agonist, on oxygen and glucose deprivation/re-oxygenation (OGD/R)-induced damage in intestinal epithelial cells (IEC-6).Terlipressin acetate exerts anti-inflammatory and anti-oxidative effects. Terlipressin acetate has the potential for hepatorenal syndrome and norepinephrine-resistant septic shock research. Purity 99.69%