Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Terlipressin diacetate | 1884420-36-3 | 99.8% | 1347.48 g/mol | C56H82N16O19S2 | 10MG
SDP

Catalog No. 5000237093
Encompass_Preferred

Terlipressin diacetate is a synthetic vasopressin analogue and selective V1 receptor agonist used in preclinical and translational research to reduce splanchnic blood flow and portal pressure. It is investigated for applications such as control of acute variceal bleeding, hepatorenal syndrome, and vasopressor-refractory septic shock.

  • Selective vasopressin V1 receptor agonist for vasoactive research.
  • Useful in studies of portal hypertension, variceal bleeding, and hepatorenal syndrome.
  • Synthetic peptide with molecular weight 1347.48 g/mol.
  • Chemical formula C56H82N16O19S2.
  • High purity (≈99.8%) suitable for research applications.
  • Supplied as a solid; recommended storage -80°C for long-term stability.

Catalog No. 50-002-37093 Supplier Medchemexpress LLC Supplier No. HY12554A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Terlipressin diacetate is a synthetic vasopressin analogue and selective V1 receptor agonist used in preclinical and translational research to reduce splanchnic blood flow and portal pressure. It is investigated for applications such as control of acute variceal bleeding, hepatorenal syndrome, and vasopressor-refractory septic shock.

  • Selective vasopressin V1 receptor agonist for vasoactive research.
  • Useful in studies of portal hypertension, variceal bleeding, and hepatorenal syndrome.
  • Synthetic peptide with molecular weight 1347.48 g/mol.
  • Chemical formula C56H82N16O19S2.
  • High purity (≈99.8%) suitable for research applications.
  • Supplied as a solid; recommended storage -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.